Learn More
Perkin Elmer US LLC Autosampler Probe for ST Nebulizers, 0.3 mm I.D.
Catalog No. 509047685
Encompass_Preferred
Replacement probe for ESI autosamplers. All perfluoroalkyl (PFA) wetted sample flow path reduces contamination and provides chemical resistance.
Catalog No. 50-904-7685 Supplier Perkin Elmer US LLC Supplier No. N0777224
Replacement probe for ESI autosamplers. All perfluoroalkyl (PFA) wetted sample flow path reduces contamination and provides chemical resistance.