Learn More
Perkin Elmer US LLC Autosampler Probe for ST Nebulizers, 0.5 mm I.D.
Catalog No. 509047686
Encompass_Preferred
135 mm- probe support length (length of carbon fiber) 80 cm- capillary length after the probe to the nebulizer connection
Catalog No. 50-904-7686 Supplier Perkin Elmer US LLC Supplier No. N0777225
135 mm- probe support length (length of carbon fiber) 80 cm- capillary length after the probe to the nebulizer connection