Learn More
Perkin Elmer US LLC ESI Capillary Tube for Spare Probe Assembly, 0.006 in
Catalog No. 502113337
Encompass_Preferred
The innovative separation probes and holders facilitate chromatography directly at the ionization source. This minimizes dead volume effects in the system for superior signal-to-noise ratios, enhanced sensitivity and narrower peaks. Compatible with LC/MS systems using Flexar SQ300 or AxION 2 TOF MS.
Catalog No. 50-211-3337 Supplier Perkin Elmer US LLC Supplier No. MZ10190317
The innovative separation probes and holders facilitate chromatography directly at the ionization source. This minimizes dead volume effects in the system for superior signal-to-noise ratios, enhanced sensitivity and narrower peaks. Compatible with LC/MS systems using Flexar SQ300 or AxION 2 TOF MS.