Learn More
AB Sciex LLC ESI Negative Calibration Solution 5600
Catalog No. NC1392787
Encompass
The calibration solution for the TripleTOF 5600 System is intended for use in negative ion mode with the DuoSpray Source. The calibration solution is introduced using the ESI probe; sample analysis is with the APCI probe.
Catalog No. NC1392787 Supplier AB Sciex LLC Supplier No. 4463277
The calibration solution for the TripleTOF 5600 System is intended for use in negative ion mode with the DuoSpray Source. The calibration solution is introduced using the ESI probe; sample analysis is with the APCI probe.