Learn More
AB Sciex LLC ESI Negative Calibration Solution for the SCIEX X500 System
Catalog No. NC1619029
Encompass
Calibration solution for the SCIEX X500 system. This solution is intended for use in ESI negative ion mode.
Catalog No. NC1619029 Supplier AB Sciex LLC Supplier No. 5042913
Calibration solution for the SCIEX X500 system. This solution is intended for use in ESI negative ion mode.