Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp ESI Probe, 200 mm x 50 µm, SFC;

Catalog No. 502039718 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

ESI Probe, 200 mm x 50 µm, SFC;

Catalog No. 50-203-9718 Supplier Waters Corp Supplier No. 700009771
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ESI Probe, 200 mm x 50 µm, SFC;

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.