Learn More
Waters Corp Waters ESI Probe Assembly, 500 mm, 100 μm
Catalog No. 502039694 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement ESI probe assembly is 500 mm in length and is used on ACQUITY QDa instruments.
  • Replacement ESI probe assembly.
  • 500 mm length.
  • 100 μm diameter.
  • Compatible with ACQUITY QDa Mass Detector and ACQUITY QDa II Mass Detector.
Catalog No. 50-203-9694 Supplier Waters Corp Supplier No. 700009642
This replacement ESI probe assembly is 500 mm in length and is used on ACQUITY QDa instruments.
  • Replacement ESI probe assembly.
  • 500 mm length.
  • 100 μm diameter.
  • Compatible with ACQUITY QDa Mass Detector and ACQUITY QDa II Mass Detector.