Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters ESI Probe Assembly, 500 mm, 125 μm

Catalog No. 502039818 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Waters ESI Probe Assembly, 500 mm, 125 μm

Catalog No. 50-203-9818 Supplier Waters Corp Supplier No. 700011241
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Waters ESI Probe Assembly, 500 mm, 125 μm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.