Learn More
Perkin Elmer US LLC ESI Probe for Spare Probe Assemblies
Catalog No. 502113336
Encompass_Preferred
The innovative separation probes and holders facilitate chromatography directly at the ionization source. This minimizes dead volume effects in the system for superior signal-to-noise ratios, enhanced sensitivity and narrower peaks. Compatible with LC/MS systems using Flexar SQ300 or AxION 2 TOF MS.
Catalog No. 50-211-3336 Supplier Perkin Elmer US LLC Supplier No. MZ109197
The innovative separation probes and holders facilitate chromatography directly at the ionization source. This minimizes dead volume effects in the system for superior signal-to-noise ratios, enhanced sensitivity and narrower peaks. Compatible with LC/MS systems using Flexar SQ300 or AxION 2 TOF MS.