Learn More
Cayman Chemical EslIcarbazepIn 10mg
SDP
Catalog No. 502913161
Encompass_Preferred
A voltage-gated sodium channel inhibitor and an active metabolite of eslicarbazepine acetate; enhances slow activation of voltage-gated sodium channels, reducing channel availability; protective against MES-induced seizures in mice (ED50 = 17.1 mg/kg)
Catalog No. 50-291-3161 Supplier Cayman Chemical Supplier No. 3500810 MG
A voltage-gated sodium channel inhibitor and an active metabolite of eslicarbazepine acetate; enhances slow activation of voltage-gated sodium channels, reducing channel availability; protective against MES-induced seizures in mice (ED50 = 17.1 mg/kg)