Learn More
Cayman Chemical Eslicarbazepine Acetate, Purity: ≥95%, CAS: 236395-14-5, 50 mg
SDP
Catalog No. 502995028
Encompass_Preferred
A sodium channel blocker (IC50 = 138 nM in a radioligand binding assay); inhibits sodium uptake in a dose-dependent manner in rat cortical synaptosomes; protective against MES-induced seizures in mice (ED50s = 4.7 and 6.3 mg/kg, for oral and i.p. administration respectively); high-dose administration (30 mg/kg) prevents picrotoxin-induced seizures; low-dose administration (10 mg/kg) reduces seizure number and duration
Catalog No. 50-299-5028 Supplier Cayman Chemical Supplier No. 2365650 MG
A sodium channel blocker (IC50 = 138 nM in a radioligand binding assay); inhibits sodium uptake in a dose-dependent manner in rat cortical synaptosomes; protective against MES-induced seizures in mice (ED50s = 4.7 and 6.3 mg/kg, for oral and i.p. administration respectively); high-dose administration (30 mg/kg) prevents picrotoxin-induced seizures; low-dose administration (10 mg/kg) reduces seizure number and duration