Learn More
Medchemexpress LLC HY-114703 10mg Medchemexpress, Eslicarbazepine CAS:104746-04-5 Purity:>98%
SDP
Catalog No. 501969369
Encompass_Preferred
Medchemexpress, HY-114703 10mg Eslicarbazepine CAS:104746-04-5 Eslicarbazepine is an oral anticonvulsant indicated for the adjunctive treatment of partial seizures. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9369 Supplier Medchemexpress LLC Supplier No. HY11470310MG
Medchemexpress, HY-114703 10mg Eslicarbazepine CAS:104746-04-5 Eslicarbazepine is an oral anticonvulsant indicated for the adjunctive treatment of partial seizures. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.