Learn More
Medchemexpress LLC HY-B0703 10mg Medchemexpress, Eslicarbazepine acetate CAS:236395-14-5 Purity:>98%
SDP
Catalog No. 502082644
Encompass_Preferred
Medchemexpress, HY-B0703 10mg Eslicarbazepine acetate CAS:236395-14-5 Eslicarbazepine acetate (BIA 2-093), an antiepileptic drug, is a dual a dual Inhibitor of β-Secretase and voltage-gated sodium channel. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2644 Supplier Medchemexpress LLC Supplier No. HYB070310MG
Medchemexpress, HY-B0703 10mg Eslicarbazepine acetate CAS:236395-14-5 Eslicarbazepine acetate (BIA 2-093), an antiepileptic drug, is a dual a dual Inhibitor of β-Secretase and voltage-gated sodium channel. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.