Learn More
Apexbio Technology LLC Eslicarbazepine acetate 236395-14-5 100mg
SDP
Catalog No. 5000096395
Encompass_Preferred
Eslicarbazepine acetate (CAS 236395-14-5) is an orally administered anticonvulsant prodrug that undergoes rapid first-pass hydrolysis to yield eslicarbazepine its pharmacologically active metabolite Eslicarbazepine predominantly modulates neuronal excitability by stabilizing the inactivated state of voltage-gated sodium channels thereby reducing sustained repetitive neuronal firing associated with seizure activity While approved for adjunctive treatment in partial-onset seizures eslicarbazepine acetate is frequently utilized in research to investigate the pathophysiology of epileptic disorders and to study sodium channel-mediated neuronal signaling
Catalog No. 50-000-96395 Supplier Apexbio Technology LLC Supplier No. B3487100
Eslicarbazepine acetate (CAS 236395-14-5) is an orally administered anticonvulsant prodrug that undergoes rapid first-pass hydrolysis to yield eslicarbazepine its pharmacologically active metabolite Eslicarbazepine predominantly modulates neuronal excitability by stabilizing the inactivated state of voltage-gated sodium channels thereby reducing sustained repetitive neuronal firing associated with seizure activity While approved for adjunctive treatment in partial-onset seizures eslicarbazepine acetate is frequently utilized in research to investigate the pathophysiology of epileptic disorders and to study sodium channel-mediated neuronal signaling