Learn More
Medchemexpress LLC Eslicarbazepine acetate | 236395-14-5 | >99.9% | 100 MG
SDP
Catalog No. 5000405565
Encompass_Preferred
Eslicarbazepine acetate is an antiepileptic agent that acts as a dual inhibitor of β-Secretase and voltage-gated sodium channels. This prodrug is activated to eslicarbazepine, an active metabolite of oxcarbazepine, sharing an identical mechanism of action. It may offer improved tolerability compared to oxcarbazepine due to potentially not producing as high peak levels of (S)-(+)-licarbazepine immediately after dosing.
  • Antiepileptic agent
  • Dual inhibitor of β-Secretase and voltage-gated sodium channel
  • Prodrug for (S)-(+)-licarbazepine
  • Purity of 99.92%
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-05565 Supplier Medchemexpress LLC Supplier No. HYB0703100MG
Eslicarbazepine acetate is an antiepileptic agent that acts as a dual inhibitor of β-Secretase and voltage-gated sodium channels. This prodrug is activated to eslicarbazepine, an active metabolite of oxcarbazepine, sharing an identical mechanism of action. It may offer improved tolerability compared to oxcarbazepine due to potentially not producing as high peak levels of (S)-(+)-licarbazepine immediately after dosing.
  • Antiepileptic agent
  • Dual inhibitor of β-Secretase and voltage-gated sodium channel
  • Prodrug for (S)-(+)-licarbazepine
  • Purity of 99.92%
  • Solid appearance
  • White to off-white color