Learn More
Selleck Chemical LLC Eslicarbazepine Acetate S4657-100mg
SDP
Catalog No. 503111543
Encompass_Preferred
Eslicarbazepine Acetate (BIA 2093 Zebinix Exalief Stedesa Aptiom) an antiepileptic drug is a dual a dual Inhibitor of -Secretase and voltage-gated sodium channel
Catalog No. 50-311-1543 Supplier Selleck Chemical LLC Supplier No. S4657100MG
Eslicarbazepine Acetate (BIA 2093 Zebinix Exalief Stedesa Aptiom) an antiepileptic drug is a dual a dual Inhibitor of -Secretase and voltage-gated sodium channel