Learn More
Enterprise Technology Solutions ESM Solar Panel,200 Watt,20Volt,9.8' Cable,31-1/2"x126",Rich BK
SDP
Supplier:  Enterprise Technology Solutions ESMDH2000601
Encompass_Preferred
Portable, 20-volt solar panel is equipped with 200 watts of peak power. Foldable, flexible design is IP65 water-resistant and includes a 9.8' universal connector cable and an angle-adjustable stand. It is the perfect companion to a DieHard portable power station (not included), giving you green power wherever you need it. Unit operates in all types of weather and withstands extreme temperatures from 14 degrees to 158 degrees Fahrenheit.
Catalog No. 50-007-39770