Learn More
Biomatik Corporation Esm1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11063
Encompass_Preferred
Esm1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ESM1. The Esm1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Endothelial Cell-Specific Molecule 1 Protein (20-184Aa). Applications: ELISA, IHC. Target Synonyms: ESM1. Accession Number: Q9NQ30. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9253