Learn More
Cayman Chemical Esmolol AcIdhydrochlorIde 10mg
SDP
Catalog No. 502913414
Encompass_Preferred
An active metabolite of esmolol; inhibits isoproterenol-induced increases in the beating rate of isolated guinea pig right atria (pA2 = 3.73); prevents isoproterenol-induced tachycardia and hypotension in anesthetized dogs; a potential impurity in commercial preparations of esmolol
Catalog No. 50-291-3414 Supplier Cayman Chemical Supplier No. 3529510 MG
An active metabolite of esmolol; inhibits isoproterenol-induced increases in the beating rate of isolated guinea pig right atria (pA2 = 3.73); prevents isoproterenol-induced tachycardia and hypotension in anesthetized dogs; a potential impurity in commercial preparations of esmolol