Learn More
Cayman Chemical Esmolol Acid (hydrochloride), Purity: ≥98%, CAS: 83356-60-9, 50 mg
SDP
Catalog No. 502913415
Encompass_Preferred
An active metabolite of esmolol; inhibits isoproterenol-induced increases in the beating rate of isolated guinea pig right atria (pA2 = 3.73); prevents isoproterenol-induced tachycardia and hypotension in anesthetized dogs; a potential impurity in commercial preparations of esmolol
Catalog No. 50-291-3415 Supplier Cayman Chemical Supplier No. 3529550 MG
An active metabolite of esmolol; inhibits isoproterenol-induced increases in the beating rate of isolated guinea pig right atria (pA2 = 3.73); prevents isoproterenol-induced tachycardia and hypotension in anesthetized dogs; a potential impurity in commercial preparations of esmolol