Learn More
Cayman Chemical Esmolol-d7 (hydrochloride), Purity: ≥99% deuterated forms (d1-d7), CAS: 1346598-13-7, 10 mg
SDP
Catalog No. 502988767
Encompass_Preferred
An internal standard for the quantification of esmolol by GC- or LC-MS
Catalog No. 50-298-8767 Supplier Cayman Chemical Supplier No. 3072710 MG
An internal standard for the quantification of esmolol by GC- or LC-MS