Learn More
Apexbio Technology LLC Esmolol HCl 81161-17-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094556
Encompass_Preferred
Esmolol HCl (CAS 81161-17-3) is a cardioselective -adrenergic receptor antagonist that attenuates sympathetic stimulation of cardiac tissue By competitively inhibiting -adrenergic receptors it reduces heart rate and myocardial contractility without significant activity at -receptors Esmolol HCl is frequently employed in research to study cardiac electrophysiology autonomic regulation and pharmacological modulation of arrhythmias Its rapid onset and short duration of action make it suitable for experimental models investigating acute cardiovascular responses or -adrenergic signaling pathways
Catalog No. 50-000-94556 Supplier Apexbio Technology LLC Supplier No. B17485.1
Esmolol HCl (CAS 81161-17-3) is a cardioselective -adrenergic receptor antagonist that attenuates sympathetic stimulation of cardiac tissue By competitively inhibiting -adrenergic receptors it reduces heart rate and myocardial contractility without significant activity at -receptors Esmolol HCl is frequently employed in research to study cardiac electrophysiology autonomic regulation and pharmacological modulation of arrhythmias Its rapid onset and short duration of action make it suitable for experimental models investigating acute cardiovascular responses or -adrenergic signaling pathways