Learn More
Apexbio Technology LLC Esmolol HCl 81161-17-3 50mg
SDP
Catalog No. 5000094654
Encompass_Preferred
Esmolol HCl (CAS 81161-17-3) is a cardioselective -adrenergic receptor antagonist that attenuates sympathetic stimulation of cardiac tissue By competitively inhibiting -adrenergic receptors it reduces heart rate and myocardial contractility without significant activity at -receptors Esmolol HCl is frequently employed in research to study cardiac electrophysiology autonomic regulation and pharmacological modulation of arrhythmias Its rapid onset and short duration of action make it suitable for experimental models investigating acute cardiovascular responses or -adrenergic signaling pathways
Catalog No. 50-000-94654 Supplier Apexbio Technology LLC Supplier No. B174850
Esmolol HCl (CAS 81161-17-3) is a cardioselective -adrenergic receptor antagonist that attenuates sympathetic stimulation of cardiac tissue By competitively inhibiting -adrenergic receptors it reduces heart rate and myocardial contractility without significant activity at -receptors Esmolol HCl is frequently employed in research to study cardiac electrophysiology autonomic regulation and pharmacological modulation of arrhythmias Its rapid onset and short duration of action make it suitable for experimental models investigating acute cardiovascular responses or -adrenergic signaling pathways