Learn More
Cayman Chemical Esmolol (hydrochloride), Purity: ≥98%, CAS: 81161-17-3, 10 mg
SDP
Catalog No. 502986703
Encompass_Preferred
A β1-AR antagonist; binds to β1-ARs (Kd = 100 nM in isolated cardiac myocytes); 34-fold selective for β1- over β2-ARs; inhibits ICa.L and the INa in rat cardiac myocytes (IC50s = 50 and 169 μM, respectively)
Catalog No. 50-298-6703 Supplier Cayman Chemical Supplier No. 2258110 MG
A β1-AR antagonist; binds to β1-ARs (Kd = 100 nM in isolated cardiac myocytes); 34-fold selective for β1- over β2-ARs; inhibits ICa.L and the INa in rat cardiac myocytes (IC50s = 50 and 169 μM, respectively)