Learn More
Cayman Chemical Esomeprazole, Purity: ≥98%, CAS: 119141-88-7, 50 mg
SDP
Catalog No. 502943739
Encompass_Preferred
An H+/K+-ATPase inhibitor; reversibly inhibits the activity of the H+/K+-ATPase in an enzyme assay; inhibits histamine-induced gastric acid secretion in rats at 30 mg/kg, s.c.
Catalog No. 50-294-3739 Supplier Cayman Chemical Supplier No. 4085150 MG
An H+/K+-ATPase inhibitor; reversibly inhibits the activity of the H+/K+-ATPase in an enzyme assay; inhibits histamine-induced gastric acid secretion in rats at 30 mg/kg, s.c.