Learn More
Apexbio Technology LLC Esomeprazole Magnesium trihydrate 217087-09-7 100mg
SDP
Catalog No. 5000094073
Encompass_Preferred
Esomeprazole Magnesium trihydrate (CAS 217087-09-7) is the S-enantiomer of omeprazole formulated as a magnesium salt It functions as a proton pump inhibitor selectively inhibiting the H /K -ATPase enzyme in gastric parietal cells to suppress gastric acid secretion This compound demonstrates pH-dependent stability degrading rapidly in acidic environments while remaining stable under alkaline conditions Controlled clinical studies have demonstrated that esomeprazole reduces the incidence of gastroduodenal ulcers by 70 85% and lowers gastrointestinal bleeding risk by up to 90% in at-risk populations It serves as a valuable tool for research into acid-related gastrointestinal disorders
Catalog No. 50-000-94073 Supplier Apexbio Technology LLC Supplier No. A3399100
Esomeprazole Magnesium trihydrate (CAS 217087-09-7) is the S-enantiomer of omeprazole formulated as a magnesium salt It functions as a proton pump inhibitor selectively inhibiting the H /K -ATPase enzyme in gastric parietal cells to suppress gastric acid secretion This compound demonstrates pH-dependent stability degrading rapidly in acidic environments while remaining stable under alkaline conditions Controlled clinical studies have demonstrated that esomeprazole reduces the incidence of gastroduodenal ulcers by 70 85% and lowers gastrointestinal bleeding risk by up to 90% in at-risk populations It serves as a valuable tool for research into acid-related gastrointestinal disorders