Learn More
Cayman Chemical Esomeprazole MagnsIum 10mg
SDP
Catalog No. 502968215
Encompass_Preferred
A form of esomeprazole in complex with magnesium; reduces aceclofenac-induced ulcer area and lesion index in the stomach in a rat model of NSAID-induced gastric ulcer at 20 mg/kg
Catalog No. 50-296-8215 Supplier Cayman Chemical Supplier No. 1732610 MG
A form of esomeprazole in complex with magnesium; reduces aceclofenac-induced ulcer area and lesion index in the stomach in a rat model of NSAID-induced gastric ulcer at 20 mg/kg