Learn More
Cayman Chemical Esomeprazole Magnesium, Purity: ≥95%, CAS: 161973-10-0, 50 mg
SDP
Catalog No. 502967861
Encompass_Preferred
A form of esomeprazole in complex with magnesium; reduces aceclofenac-induced ulcer area and lesion index in the stomach in a rat model of NSAID-induced gastric ulcer at 20 mg/kg
Catalog No. 50-296-7861 Supplier Cayman Chemical Supplier No. 1732650 MG
A form of esomeprazole in complex with magnesium; reduces aceclofenac-induced ulcer area and lesion index in the stomach in a rat model of NSAID-induced gastric ulcer at 20 mg/kg