Learn More
Medchemexpress LLC Esomeprazole ((S)-Omeprazole) | 119141-88-7 | MFCD09907604 | 99.3% | 100 MG
SDP
Catalog No. 5000385070
Encompass_Preferred
Esomeprazole ((S)-Omeprazole) is a potent and orally active proton pump inhibitor that reduces acid secretion through inhibition of the H+, K+-ATPase in gastric parietal cells. It has potential for symptomatic gastroesophageal reflux disease research.
  • Potent and orally active proton pump inhibitor
  • Reduces acid secretion by inhibiting H+, K+-ATPase in gastric parietal cells
  • Potential for symptomatic gastroesophageal reflux disease research
  • Suppresses growth of triple-negative breast cancer cells in vitro
  • Inhibits progression of fibrosis throughout the lungs and reduces circulating markers of inflammation and fibrosis
Catalog No. 50-003-85070 Supplier Medchemexpress LLC Supplier No. HY17021100MG
Esomeprazole ((S)-Omeprazole) is a potent and orally active proton pump inhibitor that reduces acid secretion through inhibition of the H+, K+-ATPase in gastric parietal cells. It has potential for symptomatic gastroesophageal reflux disease research.
  • Potent and orally active proton pump inhibitor
  • Reduces acid secretion by inhibiting H+, K+-ATPase in gastric parietal cells
  • Potential for symptomatic gastroesophageal reflux disease research
  • Suppresses growth of triple-negative breast cancer cells in vitro
  • Inhibits progression of fibrosis throughout the lungs and reduces circulating markers of inflammation and fibrosis