Learn More
Medchemexpress LLC Esomeprazole (sodium) | 161796-78-7 | 99.9% | 367.40 | 50 MG
SDP
Catalog No. 5000384665
Encompass_Preferred
Esomeprazole sodium is a potent and orally active proton pump inhibitor. It reduces acid secretion by inhibiting H+, K+-ATPase in gastric parietal cells. It also acts as an exosome inhibitor by blocking exosome release via the inhibition of V-H+-ATPases. It has potential for research related to symptomatic gastroesophageal reflux disease.
  • Potent and orally active proton pump inhibitor.
  • Reduces acid secretion by inhibiting H+, K+-ATPase in gastric parietal cells.
  • Acts as an exosome inhibitor.
  • Potential for research in symptomatic gastroesophageal reflux disease.
  • Suppresses growth of triple-negative breast cancer cells in vitro.
  • Inhibits progression of fibrosis throughout the lungs in animal models.
Catalog No. 50-003-84665 Supplier Medchemexpress LLC Supplier No. HY1702350MG
Esomeprazole sodium is a potent and orally active proton pump inhibitor. It reduces acid secretion by inhibiting H+, K+-ATPase in gastric parietal cells. It also acts as an exosome inhibitor by blocking exosome release via the inhibition of V-H+-ATPases. It has potential for research related to symptomatic gastroesophageal reflux disease.
  • Potent and orally active proton pump inhibitor.
  • Reduces acid secretion by inhibiting H+, K+-ATPase in gastric parietal cells.
  • Acts as an exosome inhibitor.
  • Potential for research in symptomatic gastroesophageal reflux disease.
  • Suppresses growth of triple-negative breast cancer cells in vitro.
  • Inhibits progression of fibrosis throughout the lungs in animal models.