Learn More
TARGETMOL CHEMICALS INC ESOMEPRAZOLE SODIUM 50MG
SDP
Catalog No. 502577062
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 250 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Esomeprazole Sodium ((S)-Omeprazole sodium) is the S-isomer of omeprazole with selective and irreversible proton pump inhibitor activity.Esomeprazole acts as an exosome inhibitor by blocking the exosome release via the inhibition of V-H+-ATPasespurity: 99%
Catalog No. 50-257-7062 Supplier TARGETMOL CHEMICALS INC Supplier No. T2686L50MG
Also available in 5 mg 10 mg 25 mg 100 mg 250 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Esomeprazole Sodium ((S)-Omeprazole sodium) is the S-isomer of omeprazole with selective and irreversible proton pump inhibitor activity.Esomeprazole acts as an exosome inhibitor by blocking the exosome release via the inhibition of V-H+-ATPasespurity: 99%