Learn More
Abcam Esomeprazole sodium, H+/ K+-ATPase (proton pump) inhibitor, 250MG
Catalog No. 502707548
Encompass_Preferred
MW 367.4 Da, Purity >99%. H+ / K+-ATPase (proton pump) inhibitor (IC50 = 2.3 µM). *S*-enantiomer of omeprazole. Attenuates intestinal mucosal barrier damage and can promote healing of gastric lesions in rats. Also inhibits Cytochrome P450 2C19 (CYP2C9), CYP2C19 and CYP3A4 (Ki values are 81.5, 8.6 and 46.6 µM, respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7548 Supplier Abcam Supplier No. AB120500250MG
MW 367.4 Da, Purity >99%. H+ / K+-ATPase (proton pump) inhibitor (IC50 = 2.3 µM). *S*-enantiomer of omeprazole. Attenuates intestinal mucosal barrier damage and can promote healing of gastric lesions in rats. Also inhibits Cytochrome P450 2C19 (CYP2C9), CYP2C19 and CYP3A4 (Ki values are 81.5, 8.6 and 46.6 µM, respectively).
The product is subject to the following: Abcam Restricted Use Statement