Learn More
Cayman Chemical EsomeprazolesodIum salt 100mg
SDP
Catalog No. 502966827
Encompass_Preferred
An H+/K+-ATPase inhibitor; reversibly inhibits the activity of the H+/K+-ATPase in an enzyme assay; inhibits histamine-induced gastric acid secretion in rats at 30 mg/kg, s.c.
Catalog No. 50-296-6827 Supplier Cayman Chemical Supplier No. 17303100 MG
An H+/K+-ATPase inhibitor; reversibly inhibits the activity of the H+/K+-ATPase in an enzyme assay; inhibits histamine-induced gastric acid secretion in rats at 30 mg/kg, s.c.