Learn More
Cayman Chemical Esomeprazole (sodium salt), Purity: ≥98%, CAS: 161796-78-7, 50 mg
SDP
Catalog No. 502965959
Encompass_Preferred
An H+/K+-ATPase inhibitor; reversibly inhibits the activity of the H+/K+-ATPase in an enzyme assay; inhibits histamine-induced gastric acid secretion in rats at 30 mg/kg, s.c.
Catalog No. 50-296-5959 Supplier Cayman Chemical Supplier No. 1730350 MG
An H+/K+-ATPase inhibitor; reversibly inhibits the activity of the H+/K+-ATPase in an enzyme assay; inhibits histamine-induced gastric acid secretion in rats at 30 mg/kg, s.c.