Learn More
Discovery Life Sciences Esophageal Cncr Mutation PBMC
SDP
Catalog No. 5000804016
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04016 Supplier Discovery Life Sciences Supplier No. 120250
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays