Learn More
Med Vet International Esophageal Foreign Body Forceps, 25cm shaft x 3.0mm diameter, J0212EFB, Each
Catalog No. 5000585483
Encompass_Preferred
One day you will need these!! An especially long forceps that can remove foreign bodies such as fish hooks or lodged bones from even the distal esophagus or trachea.Backward facing teeth for maximum gripshaft diameter small as possible to allow placement of endoscope alongside
Catalog No. 50-005-85483 Supplier Med Vet International Supplier No. JOR212EFB
One day you will need these!! An especially long forceps that can remove foreign bodies such as fish hooks or lodged bones from even the distal esophagus or trachea.Backward facing teeth for maximum gripshaft diameter small as possible to allow placement of endoscope alongside