Learn More
US Biomax Inc Esophageal and gastroesophageal junction (GEJ) adenocarcinoma and undifferentiated carcinoma with normal esophageal tissue array, including TNM, clinical stage and pathology grade,40 cases/80 cores (core size 1.5mm), replacing ES8011b
SDP
Catalog No. NC3789262 Shop All US Biomax Inc Products
Encompass
Esophageal and gastroesophageal junction (GEJ) adenocarcinoma and normal tissue microarray, containing 32 cases of adenocarcinoma, 3 undifferentiated carcinoma and 5 normal tissue, duplicated cores per case
Catalog No. NC3789262 Supplier US Biomax Inc Supplier No. ES8011C
Esophageal and gastroesophageal junction (GEJ) adenocarcinoma and normal tissue microarray, containing 32 cases of adenocarcinoma, 3 undifferentiated carcinoma and 5 normal tissue, duplicated cores per case