Learn More
New England Biolabs, Inc. Esp3I – 300 units
SDP
Catalog No. 501527863
Encompass
A Type IIS restriction endonuclease that recognizes the sequence CGTCTCN^NNNN_....
Catalog No. 50-152-7863 Supplier New England Biolabs, Inc. Supplier No. R0734S
A Type IIS restriction endonuclease that recognizes the sequence CGTCTCN^NNNN_....