Learn More
New England Biolabs, Inc. Esp3I – 1500 units
SDP
Catalog No. 501527862
Encompass
A Type IIS restriction endonuclease that recognizes the sequence CGTCTCN^NNNN_....
Catalog No. 50-152-7862 Supplier New England Biolabs, Inc. Supplier No. R0734L
A Type IIS restriction endonuclease that recognizes the sequence CGTCTCN^NNNN_....