Learn More
Sino Biological Espin Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 201806T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Espin fragment, and purified by antigen affinity chromatography.
Catalog No. 50-224-6424