Learn More
Enzo Life Sciences Estrogen receptor alpha monoclonal antibody (C-542) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISRA1010D
Encompass_Preferred
Clone: C-542. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human Estrogen Receptor α. UniProt ID: P03372. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat Hamster. Applications: ICC, IHC, IP, WB. Recommended Dilutions: Western Blot (1:500, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7669