Learn More
Enzo Life Sciences Estrogen receptor alpha monoclonal antibody (C-542) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISRA1010F
Encompass_Preferred
Clone: C-542. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human Estrogen Receptor α. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat Hamster. Applications: ICC, IHC, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7670