Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck ESR-Chex™ Controls
GSA_VASDP

Catalog No. 23046578 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
2 x 9 mL
4 x 9 mL
8 x 9 mL
12 x 9 mL
Level:
1, 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Level
23-046578 2 x 9 mL 1, 2
23-046581 12 x 9 mL 1, 2
23-046580 8 x 9 mL 1, 2
23-046579 4 x 9 mL 1, 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-046578 Supplier Streck Supplier No. 214102
Add to Cart
Add to Cart

ESR-Chex is a whole blood sed-rate control for automated and manual ESR testing methods. It is available in two levels and assayed for the most frequently used sedimentation rate tubes in conjunction with the Classical Westergren, Modified Westergren and Wintrobe methods.

  • Ready-to-use whole blood control that resembles patient sample.
  • Available in two levels: normal and abnormal.
  • Assayed for both manual and automated sed-rate testing methods.
  • 95-day open-vial stability, 365-day closed-vial stability.
  • Alerts the technologist to variables that may affect the accuracy of patient results.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance Internal quality control standards and regulatory compliance
Compatible Methods Both automated and manual ESR methods
Content And Storage 2°C to 10°C
For Use With (Equipment) Clinical chemistry analyzer
Type ESR-Chex Control
Intended Use Verify the precision and accuracy of both manual and automated ESR methods
Level 1, 2
Quantity 2 x 9 mL
Stability 95 Days Open, 12 Months Closed Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.