Learn More
Enzo Life Sciences Estrogen receptor monoclonal antibody (h-151) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISRA1000D
Encompass_Preferred
Clone: h-151. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence of human estrogen receptor, conjugated to KLH. Source: Purified from mouse ascites. Species reactivity: Human, Rat. Applications: ICC, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7667