Learn More
Aviva Systems Biology Corp ESR1 Antibody - N-terminal region (ARP37967_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37967_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ESR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ESR1. Alternate names: DKFZp686N23123, ER, ESR, ESRA, Era, NR3A1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56465