Learn More
Aviva Systems Biology Corp ESR1 Antibody - middle region (ARP45592_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ESR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ESR1. Alternate names: DKFZp686N23123, ER, ESR, ESRA, Era, NR3A1ApplicationWB. Species reactivity: Cow, Dog, Human, Pig, Rat, Sheep
Catalog No. 50-014-59055