Learn More
Aviva Systems Biology Corp Esr2 Antibody - N-terminal region (ARP37039_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37039_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Esr2. It was validated on Western Blot. Esr2,estrogen receptor 2 (beta),ER[b]; Estrb; ERbeta; Esr2,estrogen receptor beta,estrogen receptor beta,ER beta,ER-beta,OTTMUSP00000023299,OTTMUSP00000023324,oestrogen receptor beta,nuclear receptor subfamily 3 group A member 2The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: ER[b], ERbeta, EstrbApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-571