Learn More
Aviva Systems Biology Corp ESR2 Antibody - N-terminal region (ARP35863_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35863_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ESR2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ESR2. Alternate names: ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2ApplicationIHC, WB. Species reactivity: Human, Rat
Catalog No. 50-284-477