Learn More
Aviva Systems Biology Corp ESR2 Antibody - middle region (ARP32258_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32258_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ESR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ESR2. Alternate names: ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54502