Learn More
CELLTREAT Scientific Products 1900cm² ESRB Roller Bottle, Tissue Culture Treated, Printed Graduations, Non-Vented Cap, Sterile
SDP
Catalog No. NC3433420
Encompass_Preferred
"Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachmentVented or non-vented cap for open or closed systemsEasy-to-read black printed graduationsLarge knurled cap for easy gripRibbed surface of ESRB increases surface area and horizontal ribs bathe cells with media more evenly than bottles with vertical ribs"
Catalog No. NC3433420 Supplier CELLTREAT Scientific Products Supplier No. 229386
"Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachmentVented or non-vented cap for open or closed systemsEasy-to-read black printed graduationsLarge knurled cap for easy gripRibbed surface of ESRB increases surface area and horizontal ribs bathe cells with media more evenly than bottles with vertical ribs"